Learn More
AdipoGen Suc-Leu-Leu-Val-Tyr-AMC
SDP
Catalog No. 502562446
Encompass_Preferred
Chemical. CAS 94367-21-2. Formula C40H53N5O10. MW 763.9. Fluorogenic substrate for measuring the chymotrypsin-like peptidase activity of the 20S proteasome, calpains and other chymotrypsin-like proteases. Extinction 380nm. Emission 460 nm. This substrate is useful for inhibitor screening and kinetic analysis.
Catalog No. 50-256-2446 Supplier AdipoGen Supplier No. AGCP30016M001
Chemical. CAS 94367-21-2. Formula C40H53N5O10. MW 763.9. Fluorogenic substrate for measuring the chymotrypsin-like peptidase activity of the 20S proteasome, calpains and other chymotrypsin-like proteases. Extinction 380nm. Emission 460 nm. This substrate is useful for inhibitor screening and kinetic analysis.