Learn More
Medchemexpress LLC Suc-Leu-Leu-Val-Tyr-AMC | 94367-21-2 | 99.7% | 763.88 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP100210MG
Encompass_Preferred
Suc-Leu-Leu-Val-Tyr-AMC is a membrane-permeable calpain-specific fluorogenic substrate (Ex/Em = 390/480 nm). Its proteolytic hydrolysis liberates the highly fluorescent 7-amino-4-methylcoumarin (AMC) moiety. This product is intended for research use only.
  • Membrane-permeable calpain-specific fluorogenic substrate
  • Features excitation/emission wavelengths of 390/480 nm
  • Hydrolysis liberates highly fluorescent AMC moiety
  • For research use only
Catalog No. 50-004-22149