Learn More
Abcam Suc-Leu-Leu-Val-Tyr-AMC, Fluorogenic substrate of calpains and 20S proteasome, 1MG
Catalog No. 502706325
Encompass_Preferred
MW 763.9 Da, Purity >97%. Suc-Leu-Leu-Val-Tyr-AMC, Fluorogenic substrate of calpains and 20S proteasome.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6325 Supplier Abcam Supplier No. AB1421201MG
MW 763.9 Da, Purity >97%. Suc-Leu-Leu-Val-Tyr-AMC, Fluorogenic substrate of calpains and 20S proteasome.
The product is subject to the following: Abcam Restricted Use Statement