Learn More
AdipoGen Suc-Leu-Tyr-AMC
SDP
Catalog No. 502562444
Encompass_Preferred
Chemical. CAS 94367-20-1. Formula C29H33N3O8. MW 551.6. Fluorogenic substrate for measuring the chymotrypsin-like peptidase activity of the 20S proteasome. Fluorogenic substrate for calpain and papain. Peptide substrate for Ti protease from E.coli. Excitation 360nm. Emission 460nm. This substrate is useful for inhibitor screening and kinetic analysis.
Catalog No. 50-256-2444 Supplier AdipoGen Supplier No. AGCP30017M005
Chemical. CAS 94367-20-1. Formula C29H33N3O8. MW 551.6. Fluorogenic substrate for measuring the chymotrypsin-like peptidase activity of the 20S proteasome. Fluorogenic substrate for calpain and papain. Peptide substrate for Ti protease from E.coli. Excitation 360nm. Emission 460nm. This substrate is useful for inhibitor screening and kinetic analysis.