Learn More
AnaSpec SUC-LLVY-AMC, FLUOROGENIC 5MG
Catalog No. NC0353599
Encompass
Suc-LLVY-AMC, fluorogenic substrate[Suc-LLVY-AMC] ; MW 763.9; (5 mg) This 7-amino-4-methylcoumarin (AMC) labeled peptide is widely used as a fluorogenic substrate for 20S proteasome, calpains and other chymotrypsin-like proteases. Cleavage of this AMC peptide by the enzymes generates strongly fluorescent AMC that is monitored fluorimetrically at Abs/Em=353/442 nm.
Catalog No. NC0353599 Supplier AnaSpec Supplier No. AS63892
Suc-LLVY-AMC, fluorogenic substrate[Suc-LLVY-AMC] ; MW 763.9; (5 mg) This 7-amino-4-methylcoumarin (AMC) labeled peptide is widely used as a fluorogenic substrate for 20S proteasome, calpains and other chymotrypsin-like proteases. Cleavage of this AMC peptide by the enzymes generates strongly fluorescent AMC that is monitored fluorimetrically at Abs/Em=353/442 nm.