Learn More
Carbosynth LLC. Suc-Phe-Ala-Ala-Phe-pNA, 25mg, CAS# 110906-89-3, MF= C34H38N6O9
SDP
Supplier:  Carbosynth LLC. SUB3007PI25MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Application: Subtilisin and Myroilysin Substrate, salt: Trifluoroacetate Salt, one letter code: Suc-FAAF-pNA, MW: 674.72
Catalog No. 50-195-2435