Learn More
Med Vet International Dynarex Suction Unit w/Suction Cup Legs, up to 560 mmHg Vacuum Level, Each
Catalog No. 503045917
Encompass_Preferred
Dynarex Resp-02™ Suction Units are premium-grade medical devices designed to safely and effectively remove excess secretions and fluids from the airways of respiratory patients. These powerful suction units are easily portable and feature high-quality vacuum regulators and anti-vibration vacuum gauge settings to ensure quiet and efficient operation. Designed to provide vacuum levels up to 560 mmHg Fiber filter guards against bacteria and contaminants Clearly marked gauge allows for easy pressure monitoring Oil-less lubrication provides consistent functionality Includes an 800 cc disposable suction canister, tubing, and bacterial filter
Catalog No. 50-304-5917 Supplier Med Vet International Supplier No. 32300
Dynarex Resp-02™ Suction Units are premium-grade medical devices designed to safely and effectively remove excess secretions and fluids from the airways of respiratory patients. These powerful suction units are easily portable and feature high-quality vacuum regulators and anti-vibration vacuum gauge settings to ensure quiet and efficient operation. Designed to provide vacuum levels up to 560 mmHg Fiber filter guards against bacteria and contaminants Clearly marked gauge allows for easy pressure monitoring Oil-less lubrication provides consistent functionality Includes an 800 cc disposable suction canister, tubing, and bacterial filter