Learn More
Cayman Chemical Suc-YVAD-pNA 10mg
SDP
Catalog No. 502941149
Encompass_Preferred
A colorimetric caspase-1 and caspase-4 substrate; cleavage of the YVAD peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of caspase-1 and caspase-4 activity
Catalog No. 50-294-1149 Supplier Cayman Chemical Supplier No. 2813510 MG
A colorimetric caspase-1 and caspase-4 substrate; cleavage of the YVAD peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of caspase-1 and caspase-4 activity