Learn More
Cayman Chemical Suc-YVAD-pNA, Purity: ≥95%, CAS: 208264-84-0, 5 mg
SDP
Catalog No. 502937243
Encompass_Preferred
A colorimetric caspase-1 and caspase-4 substrate; cleavage of the YVAD peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of caspase-1 and caspase-4 activity
Catalog No. 50-293-7243 Supplier Cayman Chemical Supplier No. 281355 MG
A colorimetric caspase-1 and caspase-4 substrate; cleavage of the YVAD peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of caspase-1 and caspase-4 activity