Learn More
Cayman Chemical Suc-AAP-Abu-pNA (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502936688
Encompass_Preferred
A substrate for pancreatic elastase; cleaved by elastase to release p-NA, which can be quantified by colorimetric detection at 405 nm as a measure of enzyme activity
Catalog No. 50-293-6688 Supplier Cayman Chemical Supplier No. 247041 MG
A substrate for pancreatic elastase; cleaved by elastase to release p-NA, which can be quantified by colorimetric detection at 405 nm as a measure of enzyme activity