Learn More
Cayman Chemical SucAAPAbupNAtrIfluoroace 500ug
SDP
Catalog No. 502932986
Encompass_Preferred
A substrate for pancreatic elastase; cleaved by elastase to release p-NA, which can be quantified by colorimetric detection at 405 nm as a measure of enzyme activity
Catalog No. 50-293-2986 Supplier Cayman Chemical Supplier No. 24704500 UG
A substrate for pancreatic elastase; cleaved by elastase to release p-NA, which can be quantified by colorimetric detection at 405 nm as a measure of enzyme activity