Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinamic acid, 97%, Thermo Scientific™

Catalog No. AAL0653918
Change view
Click to view available options
Quantity:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0653918 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0653918 Supplier Thermo Scientific Chemicals Supplier No. L0653918

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 638-32-4
Molecular Formula C4H7NO3
Quantity 50g
MDL Number MFCD00008041
Beilstein 1754021
Synonym succinamic acid, butanoic acid, 4-amino-4-oxo, succinic acid monoamide, 3-carbamoylpropanoic acid, succinamidic acid, acmc-1b64y, 3-c-hydroxycarbonimidoyl propanoic acid
InChI Key JDVPQXZIJDEHAN-UHFFFAOYSA-N
SMILES NC(=O)CCC(O)=O
IUPAC Name 4-amino-4-oxobutanoic acid
Molecular Weight (g/mol) 117.10
PubChem CID 12522
ChEBI CHEBI:50398
Formula Weight 117.1
Percent Purity 97%
Chemical Name or Material Succinamic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-331-7
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.