Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinamide 98%, Thermo Scientific™

Catalog No. AC154521000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC154521000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC154521000 Supplier Thermo Scientific Chemicals Supplier No. 154521000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-14-5
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
InChI Key SNCZNSNPXMPCGN-UHFFFAOYSA-N
Synonym succinamide, succindiamide, succinic amide, succinic acid diamide, succinic diamide, succinic acid diimide, acmc-1bnit, ethane-1,2-dicarboxamide, aronis25142
PubChem CID 8036
IUPAC Name butanediamide
SMILES C(CC(=O)N)C(=O)N

Specifications

Color White
Infrared Spectrum Authentic
Linear Formula H2NCOCH2CH2CONH2
Quantity 100 g
Beilstein 02, 614
Merck Index 15, 8996
Formula Weight 116.12
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material Succinamide, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.