Learn More
Cayman Chemical SuccInate asy Reagent 100uL
SDP
Catalog No. 502893130
Encompass_Preferred
A transformation product of 6-PPD a biotransformation product of 6-PPD in zebrafish embryos an active metabolite of diphenylamine scavenges carbon radicals in an AIBN-induced methyl methacrylate polymerization assay has been found in tire wear particles roadway runoff and roadway-impacted creeks
Catalog No. 50-289-3130 Supplier Cayman Chemical Supplier No. 700021100 UL
A transformation product of 6-PPD a biotransformation product of 6-PPD in zebrafish embryos an active metabolite of diphenylamine scavenges carbon radicals in an AIBN-induced methyl methacrylate polymerization assay has been found in tire wear particles roadway runoff and roadway-impacted creeks