Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Succinic acid | 110-15-6 | MFCD00002789 | 100.0% | 118.09 g/mol | C4H6O4 | 10 MG
SDP

Catalog No. 5000296059
Encompass_Preferred

Succinic acid (standard) is a high-purity analytical reference material supplied as a solid for calibration, method development, and quantitative analysis in analytical and research laboratories. It is suitable for chromatography calibration, assay validation, and as a reference for food, pharmaceutical, and chemical testing.

  • High purity: 99.97%.
  • Physical form: solid suitable for weighing and preparation of standards.
  • CAS number 110-15-6 for unambiguous identification.
  • Molecular formula C4H6O4; molecular weight 118.09 g/mol.
  • Available in small pack sizes for analytical use, including 10 MG.
  • Certificate of analysis and safety data sheet available for handling and storage instructions.

Catalog No. 50-002-96059 Supplier Medchemexpress LLC Supplier No. HYN0420R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Succinic acid (standard) is a high-purity analytical reference material supplied as a solid for calibration, method development, and quantitative analysis in analytical and research laboratories. It is suitable for chromatography calibration, assay validation, and as a reference for food, pharmaceutical, and chemical testing.

  • High purity: 99.97%.
  • Physical form: solid suitable for weighing and preparation of standards.
  • CAS number 110-15-6 for unambiguous identification.
  • Molecular formula C4H6O4; molecular weight 118.09 g/mol.
  • Available in small pack sizes for analytical use, including 10 MG.
  • Certificate of analysis and safety data sheet available for handling and storage instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.