Learn More
Medchemexpress LLC Succinic acid-13C4 | 201595-67-7 | MFCD00145335 | 99.7% | 122.06 | 13C4H6O4 | 10 MG
SDP
Catalog No. 5000295840
Encompass_Preferred
Succinic acid-13C4 is a stable-isotope labeled form of succinic acid, supplied as a white to off-white solid for use as an internal standard, tracer, or reference material in analytical chemistry and metabolic studies. It offers high isotopic enrichment and is suitable for quantitative mass spectrometry and metabolic flux experiments.
  • Stable-isotope labeled (13C4) for accurate mass spectrometry quantitation.
  • High purity (99.7%) for reliable analytical results.
  • Compact 10 mg pack for low-quantity applications and method development.
  • Solid form, white to off-white, suitable for long-term storage when sealed.
  • Provided with storage guidance for solvent and bulk stability.
Catalog No. 50-002-95840 Supplier Medchemexpress LLC Supplier No. HYN0420S210MG
Succinic acid-13C4 is a stable-isotope labeled form of succinic acid, supplied as a white to off-white solid for use as an internal standard, tracer, or reference material in analytical chemistry and metabolic studies. It offers high isotopic enrichment and is suitable for quantitative mass spectrometry and metabolic flux experiments.
  • Stable-isotope labeled (13C4) for accurate mass spectrometry quantitation.
  • High purity (99.7%) for reliable analytical results.
  • Compact 10 mg pack for low-quantity applications and method development.
  • Solid form, white to off-white, suitable for long-term storage when sealed.
  • Provided with storage guidance for solvent and bulk stability.