Learn More
American Radiolabeled Chemicals Inc SUCCINIC ACID 14CU 250UC
SDP
Catalog No. 50352998
Encompass_Preferred
Succinic acid-14C(U) 250uCi in ethanol:water (1:9), 0.1 mCi/ml, 40-60 mCi/mmol, M.W. 118.1, Exclusive
Catalog No. 50-352-998 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 3593250UC
Succinic acid-14C(U) 250uCi in ethanol:water (1:9), 0.1 mCi/ml, 40-60 mCi/mmol, M.W. 118.1, Exclusive