Learn More
Pfanstiehl Inc Succinic Acid High Purity, Low Endotoxin, Low Metals, USP-NF, EP, BP, JPE. CHP, 1Kg, GMP , Multi-compendial Injectable Excipient Grade CAS 110-15-6
Catalog No. NC3315810 Shop All Pfanstiehl Products
Encompass
Pfanstiehl Succinic Acid - High Purity, Low Endotoxin, Low Metals is a multi-compendial buffer component that has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Succinic Acid meets parenteral (injectable) excipient grade standards, making it ideal for pharmaceutical, biologic, and vaccine bioprocessing and formulation. Pfanstiehl Brand Succinic Acid is compliant with the USP (United States Pharmacopoeia), EP (Ph. Eur.) (European Pharmacopoeia), BP (British Pharmacopoeia), JP (Japanese Pharmacopoeia) and ChP (Chinese Pharmacopoeia). ) Compendial, U.S.P. , cGMP , pharmaceutical grade .
Catalog No. NC3315810 Supplier Pfanstiehl Inc Supplier No. S1421KG
Pfanstiehl Succinic Acid - High Purity, Low Endotoxin, Low Metals is a multi-compendial buffer component that has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Succinic Acid meets parenteral (injectable) excipient grade standards, making it ideal for pharmaceutical, biologic, and vaccine bioprocessing and formulation. Pfanstiehl Brand Succinic Acid is compliant with the USP (United States Pharmacopoeia), EP (Ph. Eur.) (European Pharmacopoeia), BP (British Pharmacopoeia), JP (Japanese Pharmacopoeia) and ChP (Chinese Pharmacopoeia). ) Compendial, U.S.P. , cGMP , pharmaceutical grade .