Learn More
U.S. Pharmacopeia Succinic Acid, 110-15-6, MFCD00002789, 300mg
Catalog No. NC0267863
Encompass
Molecular formula C4H6O4, Molecular weight 118.09, Melting Point 370.4 °F (188 °C), Boiling Point 455 °F (235 °C), Flash point 320.0 °F (160.0 °C) (open cup), Synonyms: Butanedioic acid
Catalog No. NC0267863 Supplier U.S. Pharmacopeia Supplier No. 1623411
Molecular formula C4H6O4, Molecular weight 118.09, Melting Point 370.4 °F (188 °C), Boiling Point 455 °F (235 °C), Flash point 320.0 °F (160.0 °C) (open cup), Synonyms: Butanedioic acid