Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Acid, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18607043 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-043 500 g Poly Bottle
18-607-042 125 g Poly Bottle
18-607-044 2.5 kg Poly Bottle
18-607-046 6 x 500 g Poly Bottle
18-607-047 4 x 2.5 kg Poly Bottle
18-607-045 12 kg Poly Pail
18-607-048 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-043 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1685500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1685, 110-15-6, C4H6O4

Spectrum™ Chemical Succinic Acid, Crystal, Reagent, ACS is a white, odorless solid that is a dicarboxylic acid. Advances in industrial biotechnology that seek to displace petroleum-based chemicals in industrial applications are fueling growth in this product. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Melting Point 185°C to 191°C
CAS Max % 1
Quantity 500 g
Packaging Poly Bottle
Residue after Ignition 0.0002
Percent Purity 99%
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.