Learn More
Medchemexpress LLC Succinic acid-d4 | 14493-42-6 | 122.11 | 100 MG
SDP
Catalog No. 5000390286
Encompass_Preferred
Succinic-2,2,3,3-d4 acid is a deuterium labeled succinic acid. It is an intermediate product of the tricarboxylic acid cycle and a fermentation product of anaerobic metabolism. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90286 Supplier Medchemexpress LLC Supplier No. HYN0420S1100MG
Succinic-2,2,3,3-d4 acid is a deuterium labeled succinic acid. It is an intermediate product of the tricarboxylic acid cycle and a fermentation product of anaerobic metabolism. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.