Learn More
Medchemexpress LLC Succinic acid-d6 | 21668-90-6 | 124.13 | 100 MG
SDP
Catalog No. 5000388572
Encompass_Preferred
Succinic acid-d6 is the deuterium-labeled version of Succinic acid, an intermediate product of the tricarboxylic acid cycle and a fermentation product of anaerobic metabolism. It is intended for research use only and is not sold to patients.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity of 99.0%.
  • Appears as a solid.
  • Color is white to off-white.
Catalog No. 50-003-88572 Supplier Medchemexpress LLC Supplier No. HYN0420S100MG
Succinic acid-d6 is the deuterium-labeled version of Succinic acid, an intermediate product of the tricarboxylic acid cycle and a fermentation product of anaerobic metabolism. It is intended for research use only and is not sold to patients.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity of 99.0%.
  • Appears as a solid.
  • Color is white to off-white.