Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Succinic acid-d6 | 21668-90-6 | MFCD00084128 | 99.0% | 124.13 g/mol | C4D6O4 | 10 MG
SDP

Catalog No. 5000295622
Encompass_Preferred

Succinic acid-d6 is the deuterium-labeled form of succinic acid provided as a high-purity solid for use as an internal standard and tracer in analytical chemistry and metabolic studies. It enables improved quantitation and selectivity in isotope-based assays.

  • Deuterium-labeled internal standard for quantitative analysis
  • High isotopic purity (about 99.0%) for reliable trace quantitation
  • Molecular formula C4D6O4; molecular weight 124.13 g/mol
  • Solid powder, minimizes handling variability
  • Available in small mass packages suitable for analytical workflows
  • Storage: store sealed away from moisture; in solvent: -80°C (6 months), -20°C (1 month)
  • Compatible with NMR, GC-MS, and LC-MS methods

Catalog No. 50-002-95622 Supplier Medchemexpress LLC Supplier No. HYN0420S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Succinic acid-d6 is the deuterium-labeled form of succinic acid provided as a high-purity solid for use as an internal standard and tracer in analytical chemistry and metabolic studies. It enables improved quantitation and selectivity in isotope-based assays.

  • Deuterium-labeled internal standard for quantitative analysis
  • High isotopic purity (about 99.0%) for reliable trace quantitation
  • Molecular formula C4D6O4; molecular weight 124.13 g/mol
  • Solid powder, minimizes handling variability
  • Available in small mass packages suitable for analytical workflows
  • Storage: store sealed away from moisture; in solvent: -80°C (6 months), -20°C (1 month)
  • Compatible with NMR, GC-MS, and LC-MS methods

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.