Learn More
Description
- Succinic acid disodium salt is used to prepare buffer solution for the quantitative analysis of auxins.it can be also used:
- To study the effect of liquid phase on the basic properties of α-tricalcuim phosphate (α-tcp) based cement, biopex
- In the synthesis of cadmium sulfide (cds) nanoparticles using thiourea as a source for sulfide
Specifications
Specifications
| CAS | 150-90-3 |
| Molecular Formula | C4H4Na2O4 |
| Linear Formula | NaOOCCH2CH2COONa |
| MDL Number | MFCD00002790 |
| Quantity | 500 g |
| Synonym | Sodium succinate dibasic |
| Molecular Weight (g/mol) | 162.05 |
| Percent Purity | 99% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.