Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Acid Disodium Hexahydrate, MP Biomedicals™

Catalog No. MP210292991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210292991 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210292991 Supplier MP Biomedicals, Inc Supplier No. 0210292991
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use as a TCA (Krebs cycle) supplement in cell culture applications. Succinic Acid Disodium Hexahydrate, MP Biomedicals (aka, sodium succinate dibasic hexahydrate) can be used to study x-ray crystallography, protein structural analysis, proteomics and opti

  • Study x-ray crystallography, protein structural analysis, proteomics and optimization reagents
  • Determine the calorimetric and diffractometric evidence for the sequential crystallization of buffer components and the consequential pH swing in frozen solutions
  • Provide insights into the mechanism of Pseudomonas dacunhae aspartate β-decarboxylase from rapid-scanning stopped-flow kinetics
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.142
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
Synonym disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
PubChem CID 3083938
ChEBI CHEBI:63686
IUPAC Name disodium;butanedioate;hexahydrate
SMILES C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]

Specifications

Quantity 1 kg
Physical Form Crystalline Powder
Solubility Information Soluble in water (100mg/mL, clear and colorless solution).
Formula Weight 270.1
Percent Purity ≥98%
Recommended Storage Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.