Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic acid disodium salt, for biochemistry, anhydrous

Catalog No. ac459631000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459631000 100 g
AC459635000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459631000 Supplier Thermo Scientific Chemicals Supplier No. 459631000
Only null left
Add to Cart

CAS: 150-90-3 | C4H4Na2O4 | 162.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
Synonym disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277
PubChem CID 9020
ChEBI CHEBI:63675
IUPAC Name disodium;butanedioate
SMILES C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >300.0°C
pH 6 to 9 (0.4 mol sol)
Loss on Drying 0.3% max.
Infrared Spectrum Authentic
Linear Formula NaO2CCH2CH2CO2Na
Quantity 100 g
Solubility Information Solubility in water: 300g/L (25°C).
Formula Weight 162.05
Grade Biochemical
Chemical Name or Material Succinic acid disodium salt
Hazard Category Warning
Hazard Statement GHS H StatementCauses skin irritation.Causes serious eye irritation.May cause respiratory irritation.
Precautionary Statement GHS P StatementIF ON SKIN: Wash with plenty of soap and water.IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON CENTER or doctor/physician if you feel unwell.Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.