Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Acid, FCC, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607049 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-049 125 g Poly Bottle
18-607-052 12 kg Poly Pail
18-607-053 25 kg Fiber Drum
18-607-054 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-049 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1686125GM
Add to Cart
Add to Cart

S1686, 110-15-6, C4H6O4

Spectrum™ Chemical Succinic Acid, FCC is used as a flavoring agent for food and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

CAS Max % 1
Percent Purity 99.0 to 100.5%
Grade FCC
Quantity 125 g
Ignition Residue 0.00025
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.