Learn More
U.S. Pharmacopeia Succinic Acid Melting Point Standard, 110-15-6, MFCD00002789, 1g
Catalog No. NC1644247
Encompass
Molecular formula C4H6O4, Molecular weight 118.09, Melting point/freezing point 363.2 - 366.8 °F (184 - 186 °C), Initial boiling point 455 °F (235 °C), pH in aqueous solution 2.7 Solution: 0.1%
Catalog No. NC1644247 Supplier U.S. Pharmacopeia Supplier No. 1623422
Molecular formula C4H6O4, Molecular weight 118.09, Melting point/freezing point 363.2 - 366.8 °F (184 - 186 °C), Initial boiling point 455 °F (235 °C), pH in aqueous solution 2.7 Solution: 0.1%