Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Acid, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111012449 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2449 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2449 Supplier MilliporeSigma Supelco Supplier No. PHR14181G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Succinic acid is a dicarboxylic acid, which is used primarily as an acidity regulator in the food and beverage industry. Pharmaceutically, it is usually used as a conjugate feeder acid to form succinate salt and as an excipient. Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 110-15-6
Physical Form Neat
Linear Formula HOOCCH2CH2COOH
MDL Number MFCD00002789
Melting Point 184°C to 186°C
Boiling Point 235°C
Synonym Succinic acid; Butanedioic acid
Quantity 1 g
Formula Weight 118.09
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.