Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Acid, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02004326 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-326 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-326 Supplier Avantor Supplier No. 034601

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Formula Weight 118.09
Grade BAKER ANALYZED™ ACS Reagent
Quantity 500 g
Packaging Poly Bottle
Physical Form Powder
Specific Gravity 1L = 1.57kg
Chemical Name or Material Succinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.