Learn More
Medchemexpress LLC Succinic acid (standard) | 110-15-6 | 118.09 | 50 MG
SDP
Catalog No. 5000388821
Encompass_Preferred
Succinic acid (Standard) is the analytical standard of Succinic acid. This product is intended for research and analytical applications. Succinic acid is an anti-anxiety agent with oral activity. Disodium succinate is the salt form of Succinic acid. Succinic acid is an intermediate product of the tricarboxylic acid cycle. Succinic acid is an important platform chemical. Succinic acid can be used as surfactant, additive, ion chelating agent, flavoring agent and other applications in chemical, pharmaceutical and food fields.
  • Endogenous metabolite
  • Oral activity
  • Intermediate product of the tricarboxylic acid cycle
  • Important platform chemical
  • Can be used as surfactant, additive, ion chelating agent, flavoring agent in chemical, pharmaceutical, and food fields
  • Intended for research and analytical applications
Catalog No. 50-003-88821 Supplier Medchemexpress LLC Supplier No. HYN0420R50MG
Succinic acid (Standard) is the analytical standard of Succinic acid. This product is intended for research and analytical applications. Succinic acid is an anti-anxiety agent with oral activity. Disodium succinate is the salt form of Succinic acid. Succinic acid is an intermediate product of the tricarboxylic acid cycle. Succinic acid is an important platform chemical. Succinic acid can be used as surfactant, additive, ion chelating agent, flavoring agent and other applications in chemical, pharmaceutical and food fields.
  • Endogenous metabolite
  • Oral activity
  • Intermediate product of the tricarboxylic acid cycle
  • Important platform chemical
  • Can be used as surfactant, additive, ion chelating agent, flavoring agent in chemical, pharmaceutical, and food fields
  • Intended for research and analytical applications