Learn More
Medchemexpress LLC Succinic anhydride | 108-30-5 | 100.0% | 100.07 g/mol | C4H4O3 | 1 KG
SDP
Catalog No. 5000202366
Encompass_Preferred
Succinic anhydride is a cyclic anhydride used as an acylating reagent and as a building block for linker synthesis in organic and bioconjugation chemistry. Supplied as a solid reagent in a 1 KG unit, it is provided with analytical documentation for research use.
  • High purity reagent for research applications.
  • Molecular formula C4H4O3 and molecular weight 100.07 g/mol.
  • Used as an acylating reagent and ADC linker precursor.
  • Provided with certificate of analysis and safety data sheet documentation.
  • Available as a 1 KG pack for larger-scale laboratory use.
Catalog No. 50-002-02366 Supplier Medchemexpress LLC Supplier No. HY793691KG
Succinic anhydride is a cyclic anhydride used as an acylating reagent and as a building block for linker synthesis in organic and bioconjugation chemistry. Supplied as a solid reagent in a 1 KG unit, it is provided with analytical documentation for research use.
  • High purity reagent for research applications.
  • Molecular formula C4H4O3 and molecular weight 100.07 g/mol.
  • Used as an acylating reagent and ADC linker precursor.
  • Provided with certificate of analysis and safety data sheet documentation.
  • Available as a 1 KG pack for larger-scale laboratory use.