Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Succinic anhydride | 108-30-5 | MFCD00005525 | 100.0% | 100.07 | C4H4O3 | 500 G
SDP

Catalog No. 5000204105
Encompass_Preferred

Succinic anhydride is a cyclic anhydride used as an acylation reagent and a non-cleavable ADC linker intermediate for research applications. It is supplied as a solid and should be stored sealed and protected from moisture.

  • Used for organic synthesis and ADC linker formation.
  • Cas number: 108-30-5.
  • Molecular formula: C4H4O3.
  • Molecular weight: 100.07.
  • Purity: 99.96% (as listed by supplier).
  • Available as a 500 g pack.
  • Storage: 4°C sealed; in solution, -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-04105 Supplier Medchemexpress LLC Supplier No. HY79369500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Succinic anhydride is a cyclic anhydride used as an acylation reagent and a non-cleavable ADC linker intermediate for research applications. It is supplied as a solid and should be stored sealed and protected from moisture.

  • Used for organic synthesis and ADC linker formation.
  • Cas number: 108-30-5.
  • Molecular formula: C4H4O3.
  • Molecular weight: 100.07.
  • Purity: 99.96% (as listed by supplier).
  • Available as a 500 g pack.
  • Storage: 4°C sealed; in solution, -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.