Learn More
Medchemexpress LLC Succinic anhydride | 108-30-5 | MFCD00005525 | 100.0% | 100.07 g/mol | C4H4O3 | 100 G
SDP
Catalog No. 5000202365
Encompass_Preferred
Succinic anhydride is a cyclic acid anhydride supplied as a research reagent and analytical standard, commonly used in organic synthesis and bioconjugation applications. It is provided with a Certificate of Analysis and Safety Data Sheet for handling and quality information.
  • used as a reagent and analytical standard in organic synthesis.
  • commonly applied in bioconjugation and linker chemistry.
  • supplied with COA and SDS for quality and safety verification.
  • available in multiple pack sizes to support small- and bulk-scale work.
  • solid at room temperature with a known molecular weight and formula for calculations.
Catalog No. 50-002-02365 Supplier Medchemexpress LLC Supplier No. HY79369100G
Succinic anhydride is a cyclic acid anhydride supplied as a research reagent and analytical standard, commonly used in organic synthesis and bioconjugation applications. It is provided with a Certificate of Analysis and Safety Data Sheet for handling and quality information.
  • used as a reagent and analytical standard in organic synthesis.
  • commonly applied in bioconjugation and linker chemistry.
  • supplied with COA and SDS for quality and safety verification.
  • available in multiple pack sizes to support small- and bulk-scale work.
  • solid at room temperature with a known molecular weight and formula for calculations.