Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Succinic anhydride | 108-30-5 | MFCD00005525 | 100.0% | 100.07 g/mol | C4H4O3 | 50 G
SDP

Catalog No. 5000202168
Encompass_Preferred

Succinic anhydride is a cyclic dicarboxylic anhydride used as a reagent and linker intermediate in organic synthesis and bioconjugation. Supplied for laboratory use; consult the SDS and COA for handling, storage, and analytical details.

  • High purity (manufacturer-stated 99.96%).
  • CAS number 108-30-5.
  • Molecular formula C4H4O3; molecular weight 100.07 g/mol.
  • Used in derivatization, acylation, and ADC linker chemistry.
  • Available in multiple pack sizes, including 50 g.
  • SDS and COA available for safety and quality documentation.

Catalog No. 50-002-02168 Supplier Medchemexpress LLC Supplier No. HY7936950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Succinic anhydride is a cyclic dicarboxylic anhydride used as a reagent and linker intermediate in organic synthesis and bioconjugation. Supplied for laboratory use; consult the SDS and COA for handling, storage, and analytical details.

  • High purity (manufacturer-stated 99.96%).
  • CAS number 108-30-5.
  • Molecular formula C4H4O3; molecular weight 100.07 g/mol.
  • Used in derivatization, acylation, and ADC linker chemistry.
  • Available in multiple pack sizes, including 50 g.
  • SDS and COA available for safety and quality documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.