Learn More
Medchemexpress LLC Succinimide-15N | 32807-36-6 | 99.0% | 100.08 | 25 MG
SDP
Catalog No. 5000402277
Encompass_Preferred
Succinimide-15N is the 15N labeled Succinimide. Succinimide rapidly hydrolyzes into aspartic acid and aspartic acid during the reduction/alkylation process. It is utilized in the development of antiepileptic agents.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-02277 Supplier Medchemexpress LLC Supplier No. 25MGHY41877S
Succinimide-15N is the 15N labeled Succinimide. Succinimide rapidly hydrolyzes into aspartic acid and aspartic acid during the reduction/alkylation process. It is utilized in the development of antiepileptic agents.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.