Learn More
Medchemexpress LLC Succinimide (pyrrolidine-2,5-dione) | 123-56-8 | 99.9% | 99.09 | 100 G
SDP
Catalog No. 5000403479
Encompass_Preferred
Succinimide (pyrrolidine-2,5-dione) rapidly hydrolyzes into aspartic acid during the reduction/alkylation process. It is utilized in the development of an antiepileptic agent.
  • Solid appearance
  • White to off-white color
  • Used in antiepileptic agent development
Catalog No. 50-004-03479 Supplier Medchemexpress LLC Supplier No. HY41877100G
Succinimide (pyrrolidine-2,5-dione) rapidly hydrolyzes into aspartic acid during the reduction/alkylation process. It is utilized in the development of an antiepileptic agent.
  • Solid appearance
  • White to off-white color
  • Used in antiepileptic agent development