Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Succinimide (pyrrolidine-2,5-dione) | 123-56-8 | 99.9% | 99.09 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHY41877

Encompass_Preferred

Succinimide rapidly hydrolyzes into aspartic acid during the reduction/alkylation process. It is utilized in the development of antiepileptic agents.

  • High purity: available at 99.92% purity.
  • Research use: intended for research purposes.
  • Solubility: soluble in DMSO (100 mg/mL), with specific protocols for in vivo dissolution using co-solvents like PEG300, Tween-80, and Saline, or SBE-β-CD in Saline, or Corn Oil.
  • Stability: powder form is stable for 3 years at -20°C and 2 years at 4°C. In solvent, it's stable for 6 months at -80°C and 1 month at -20°C.

Catalog No. 50-004-03139


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.