Learn More
Medchemexpress LLC Succinimidyl 4-formylbenzoate | 60444-78-2 | MFCD00133733 | 98.6% | 247.20 | C12H9NO5 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY130107500MG
Encompass_Preferred
Ald-Ph-NHS ester (N-succinimidyl 4-formylbenzoate) is an NHS ester linker containing a benzaldehyde functional group used as a non-cleavable linker for antibody-drug conjugation (ADC). It reacts with primary amines to form stable amide bonds, enabling covalent attachment of payloads to antibodies or other amine-containing biomolecules. Supplied as a solid for research use.
  • Non-cleavable linker suitable for ADC development.
  • Reacts with primary amines to form stable amide bonds.
  • Benzaldehyde functional group enables further chemoselective modification.
  • Off-white to pale yellow solid, moisture sensitive.
  • Available in multiple laboratory pack sizes for research use.
Catalog No. 50-002-02009