Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate | 64987-85-5 | MFCD00009634 | 99.6% | 334.32 g/mol | C16H18N2O6 | 25 G
SDP

Catalog No. 5000202318
Encompass_Preferred

SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate) is a heterobifunctional, non-cleavable crosslinker that reacts with primary amines and sulfhydryls to form stable thioether bonds. It is supplied as an off-white to light brown solid for research use and is commonly used in protein conjugation, antibody-drug conjugate linker chemistry, and preparation of antigen-coupled splenic cells to induce antigen-specific immune responses.

  • Molecular formula: C16H18N2O6.
  • Molecular weight: 334.32 g/mol.
  • Purity: 99.58% as supplied.
  • CAS number: 64987-85-5.
  • Physical form: solid, off-white to light brown.
  • Storage: store sealed, away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Applications: protein crosslinking, ADC linker chemistry, conjugation to KLH or OVA, preparation of antigen-coupled splenic cells.

Catalog No. 50-002-02318 Supplier Medchemexpress LLC Supplier No. HY4236025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate) is a heterobifunctional, non-cleavable crosslinker that reacts with primary amines and sulfhydryls to form stable thioether bonds. It is supplied as an off-white to light brown solid for research use and is commonly used in protein conjugation, antibody-drug conjugate linker chemistry, and preparation of antigen-coupled splenic cells to induce antigen-specific immune responses.

  • Molecular formula: C16H18N2O6.
  • Molecular weight: 334.32 g/mol.
  • Purity: 99.58% as supplied.
  • CAS number: 64987-85-5.
  • Physical form: solid, off-white to light brown.
  • Storage: store sealed, away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Applications: protein crosslinking, ADC linker chemistry, conjugation to KLH or OVA, preparation of antigen-coupled splenic cells.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.