Learn More
Medchemexpress LLC Succinimidyl 6-[3-(2-pyridyldithio)propionamido]hexanoate | 158913-22-5 | MFCD00083163 | 98.1% | 425.52 | C18H23N3O5S2 | 100MG
SDP
Catalog No. 5000235572
Encompass_Preferred
SPDP-C6-NHS ester is a heterobifunctional alkyl/ether-based linker bearing an N-hydroxysuccinimide (NHS) ester and a pyridyldithiol moiety, designed for bioconjugation and assembly of bifunctional molecules such as PROTACs. The NHS ester enables amine coupling to form stable amide bonds, while the pyridyldithiol group allows disulfide formation with thiols, providing modular chemistry for linking ligands, payloads, or proteins.
Catalog No. 50-002-35572 Supplier Medchemexpress LLC Supplier No. HY124377100MG
SPDP-C6-NHS ester is a heterobifunctional alkyl/ether-based linker bearing an N-hydroxysuccinimide (NHS) ester and a pyridyldithiol moiety, designed for bioconjugation and assembly of bifunctional molecules such as PROTACs. The NHS ester enables amine coupling to form stable amide bonds, while the pyridyldithiol group allows disulfide formation with thiols, providing modular chemistry for linking ligands, payloads, or proteins.