Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Succinimidyl 6-[3-(2-pyridyldithio)propionamido]hexanoate | 158913-22-5 | MFCD00083163 | 98.1% | 425.52 | C18H23N3O5S2 | 500MG
SDP

Catalog No. 5000238859
Encompass_Preferred

SPDP-C6-NHS ester is an alkyl/ether-based PROTAC linker that features an NHS-activated ester for amine coupling and a pyridyldisulfide moiety for disulfide conjugation. Supplied as a white to off-white solid, it is used in conjugation chemistry and the synthesis of PROTACs and related bifunctional molecules.

  • Alkyl/ether-based PROTAC linker with an NHS activated ester.
  • Contains a pyridyldisulfide group for disulfide bond formation.
  • High purity: 98.1%.
  • Molecular formula C18H23N3O5S2; molecular weight 425.52.
  • Soluble in DMSO at 50 mg/mL; ultrasonic treatment recommended.
  • Store powder at -20°C for long term; store solutions at -80°C for short term.
  • Available in small-scale solid quantities for research use.

Catalog No. 50-002-38859 Supplier Medchemexpress LLC Supplier No. HY124377500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SPDP-C6-NHS ester is an alkyl/ether-based PROTAC linker that features an NHS-activated ester for amine coupling and a pyridyldisulfide moiety for disulfide conjugation. Supplied as a white to off-white solid, it is used in conjugation chemistry and the synthesis of PROTACs and related bifunctional molecules.

  • Alkyl/ether-based PROTAC linker with an NHS activated ester.
  • Contains a pyridyldisulfide group for disulfide bond formation.
  • High purity: 98.1%.
  • Molecular formula C18H23N3O5S2; molecular weight 425.52.
  • Soluble in DMSO at 50 mg/mL; ultrasonic treatment recommended.
  • Store powder at -20°C for long term; store solutions at -80°C for short term.
  • Available in small-scale solid quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.