Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Succinimidyl 6-[3-(2-pyridyldithio)propionamido]hexanoate | 158913-22-5 | MFCD00083163 | 98.1% | 425.52 g/mol | C18H23N3O5S2 | 1G
SDP

Catalog No. 5000238902
Encompass_Preferred

Succinimidyl 6-[3-(2-pyridyldithio)propionamido]hexanoate is an alkyl/ether-based PROTAC linker containing an N-hydroxysuccinimide (NHS) ester for amine coupling and a pyridyldisulfide moiety for thiol-directed conjugation. It is used in PROTAC and bioconjugation synthesis and features a reducible disulfide that enables controlled cleavage in downstream applications.

  • Provides an NHS ester for efficient conjugation to primary amines.
  • Contains a pyridyldisulfide cleavable disulfide for thiol chemistry.
  • Suitable as a linker in PROTAC and bioconjugation synthesis.
  • Molecular formula C18H23N3O5S2; molecular weight 425.52 g/mol.
  • Soluble in DMSO at approximately 50 mg/mL; may require sonication.
  • Store powder at -20°C for long-term stability; solvent solutions require low-temperature storage.
  • High purity (98.1%) suitable for research applications.

Catalog No. 50-002-38902 Supplier Medchemexpress LLC Supplier No. HY1243771G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Succinimidyl 6-[3-(2-pyridyldithio)propionamido]hexanoate is an alkyl/ether-based PROTAC linker containing an N-hydroxysuccinimide (NHS) ester for amine coupling and a pyridyldisulfide moiety for thiol-directed conjugation. It is used in PROTAC and bioconjugation synthesis and features a reducible disulfide that enables controlled cleavage in downstream applications.

  • Provides an NHS ester for efficient conjugation to primary amines.
  • Contains a pyridyldisulfide cleavable disulfide for thiol chemistry.
  • Suitable as a linker in PROTAC and bioconjugation synthesis.
  • Molecular formula C18H23N3O5S2; molecular weight 425.52 g/mol.
  • Soluble in DMSO at approximately 50 mg/mL; may require sonication.
  • Store powder at -20°C for long-term stability; solvent solutions require low-temperature storage.
  • High purity (98.1%) suitable for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.