Learn More
CPC Scientific Suc-Asp-Phe-NMePhe-Gly-Leu-Met-NH2 (trifluoroacetate salt) 5MG
SDP
Catalog No. 502110454
Encompass
SEQUENCE: Suc-Asp-Phe-NMePhe-Gly-Leu-Met-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Suc-DF-(NMeF)-GLM-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT: 841.98STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: Succinyl-(Asp,N-Me-Phe)-Substance P (6-11), Suc-Asp-Phe-N-Me-Phe-Gly-Leu-Met-NH, SenktideRESEARCH AREA: NeuropeptidesREFERENCES:Ciccocioppo, R. et al. Peptides 16, 533 (1995)Saigo, A. et al. Neurosci. Lett. 159, 187 (1993)SKU(s): SUBP-008A, SUBP-008B
Catalog No. 50-211-0454 Supplier CPC Scientific Supplier No. SUBP008B
SEQUENCE: Suc-Asp-Phe-NMePhe-Gly-Leu-Met-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Suc-DF-(NMeF)-GLM-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT: 841.98STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: Succinyl-(Asp,N-Me-Phe)-Substance P (6-11), Suc-Asp-Phe-N-Me-Phe-Gly-Leu-Met-NH, SenktideRESEARCH AREA: NeuropeptidesREFERENCES:Ciccocioppo, R. et al. Peptides 16, 533 (1995)Saigo, A. et al. Neurosci. Lett. 159, 187 (1993)SKU(s): SUBP-008A, SUBP-008B