Learn More
Medchemexpress LLC Succinyl-Coenzyme A sodium | 108347-97-3 | ≥99.0% | 1 MG
SDP
Catalog No. 5000349642
Encompass_Preferred
Succinyl-Coenzyme A sodium is a pivotal intermediate metabolite in the tricarboxylic acid cycle and a key coenzyme A metabolite. It is biosynthesized from α-ketoglutarate or propionyl-CoA. Succinyl-Coenzyme A sodium acts as a critical precursor and substrate for heme biosynthesis and gluconeogenesis. Insufficiency of Succinyl-Coenzyme A sodium caused by cobalamin deficiency is directly linked to growth retardation, impaired heme synthesis, tissue glycine accumulation, and neurological abnormalities.
  • It can be used in research on metabolic, neurological, and hematological abnormalities (such as porphyria) caused by nutritional vitamin B12 deficiency (leading to a lack of Succinyl-Coenzyme A synthesis).
  • It acts as a substrate for rat liver δ-aminolevulinic acid synthase-1 with a Michaelis constant (Km) of 6-120 μmol/L.
  • It does not bind to recombinant NLRX1 in an in vitro pull-down assay.
Catalog No. 50-003-49642 Supplier Medchemexpress LLC Supplier No. HY1378081MG
Succinyl-Coenzyme A sodium is a pivotal intermediate metabolite in the tricarboxylic acid cycle and a key coenzyme A metabolite. It is biosynthesized from α-ketoglutarate or propionyl-CoA. Succinyl-Coenzyme A sodium acts as a critical precursor and substrate for heme biosynthesis and gluconeogenesis. Insufficiency of Succinyl-Coenzyme A sodium caused by cobalamin deficiency is directly linked to growth retardation, impaired heme synthesis, tissue glycine accumulation, and neurological abnormalities.
  • It can be used in research on metabolic, neurological, and hematological abnormalities (such as porphyria) caused by nutritional vitamin B12 deficiency (leading to a lack of Succinyl-Coenzyme A synthesis).
  • It acts as a substrate for rat liver δ-aminolevulinic acid synthase-1 with a Michaelis constant (Km) of 6-120 μmol/L.
  • It does not bind to recombinant NLRX1 in an in vitro pull-down assay.