Learn More
Cayman Chemical Succinyl-Coenzyme A (sodium salt), Purity: ≥70%, CAS: 108347-97-3, 10 mg
SDP
Catalog No. 502993766
Encompass_Preferred
An intermediate in the citric acid cycle; converted to succinate; required for the first step of porphyrin and heme synthesis; deficiency can lead to neuromotor dysfunction
Catalog No. 50-299-3766 Supplier Cayman Chemical Supplier No. 2329710 MG
An intermediate in the citric acid cycle; converted to succinate; required for the first step of porphyrin and heme synthesis; deficiency can lead to neuromotor dysfunction