Learn More
TARGETMOL CHEMICALS INC SUCCINYL PHOSPHONATE TRI 5MG
SDP
Catalog No. 502581387
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Succinyl phosphonate trisodium salt is an inhibitor of (alpha)-ketoglutarate dehydrogenase (KGDHC)purity: 98%
Catalog No. 50-258-1387 Supplier TARGETMOL CHEMICALS INC Supplier No. T130195MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Succinyl phosphonate trisodium salt is an inhibitor of (alpha)-ketoglutarate dehydrogenase (KGDHC)purity: 98%