Learn More
Vector Laboratories WFA, WFL, Unconjugated, 5 mg B-1025S-5
SDP
Supplier:  Vector Laboratories B1025S5
Encompass
Wisteria Floribunda Lectin (WFA, WFL), Unconjugated, 5 mg. The binding specificity of Wisteria floribunda lectin (WFL) is not completely clear, but this lectin appears to preferentially bind carbohydrate structures terminating in N-acetylgalactosamine linked alpha or beta to the 3 or 6 position of galactose. This lectin has been used to fractionate lymphocyte populations and, though not mitogenic, elicits the production of lymphokines from murine splenocytes.
Catalog No. 50-204-5610