Learn More
Vector Laboratories Succinylated Wheat Germ Agglutinin (WGA), Fluorescein, 5mg
SDP
Catalog No. 502045652
Encompass
This derivative has been reported to have properties distinct from the native lectin. Evidence suggests that Succinylated Wheat Germ agglutinin does not bind to sialic acid residues, unlike the native form, but retains its specificity toward N-acetylglucosamine.
Catalog No. 50-204-5652 Supplier Vector Laboratories Supplier No. FL1021S5
Add to Cart
Add to Cart
This derivative has been reported to have properties distinct from the native lectin. Evidence suggests that Succinylated Wheat Germ agglutinin does not bind to sialic acid residues, unlike the native form, but retains its specificity toward N-acetylglucosamine.