Learn More
Cayman Chemical Succinylcholine (chloride hydrate), Purity: ≥95%, CAS: 6101-15-1, 25 g
SDP
Catalog No. 502948663
Encompass_Preferred
An agonist of muscle-type nAChRs (EC50 = 10.8 µM in X. laevis oocytes expressing the human receptor); selective for the muscle-type nAChR over α3β2, α3β4, α4β2, or α7 subunit-containing neuronal nAChRs (IC50s = >100 µM); induces neuromuscular block in phrenic nerve-stimulated isolated rat phrenic nerve-diaphragm preparations at 600 µg
Catalog No. 50-294-8663 Supplier Cayman Chemical Supplier No. 3282925 G
An agonist of muscle-type nAChRs (EC50 = 10.8 µM in X. laevis oocytes expressing the human receptor); selective for the muscle-type nAChR over α3β2, α3β4, α4β2, or α7 subunit-containing neuronal nAChRs (IC50s = >100 µM); induces neuromuscular block in phrenic nerve-stimulated isolated rat phrenic nerve-diaphragm preparations at 600 µg